Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (2-Bromoethyl)triphenylphosphonium bromide, 98%

Click to view available options
Quantity:
5g
25g
100g
7301-93-1
C20H19Br2P
450.154
MFCD04039352
ZHLVWYZWQPNQDQ-UHFFFAOYSA-M
2-bromoethyl triphenylphosphonium bromide,phosphonium, 2-bromoethyl triphenyl-, bromide,2-bromoethyl triphenylphosohonium bromide,2-bromoethyl triphenylphosphanium bromide,2-bromoethyl triphenyl phosphanium bromide,2-bromoethyl triphenyl phosphonium bromide,phosphonium, 2-bromoethyl triphenyl-,bromide
22210408
2-bromoethyl(triphenyl)phosphanium;bromide
C1=CC=C(C=C1)[P+](CCBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
7301-93-1
C20H19Br2P
450.154
MFCD04039352
ZHLVWYZWQPNQDQ-UHFFFAOYSA-M
2-bromoethyl triphenylphosphonium bromide,phosphonium, 2-bromoethyl triphenyl-, bromide,2-bromoethyl triphenylphosohonium bromide,2-bromoethyl triphenylphosphanium bromide,2-bromoethyl triphenyl phosphanium bromide,2-bromoethyl triphenyl phosphonium bromide,phosphonium, 2-bromoethyl triphenyl-,bromide
22210408
2-bromoethyl(triphenyl)phosphanium;bromide
C1=CC=C(C=C1)[P+](CCBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
Applications
(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Melting Point 186°C to 189°C
Quantity 100g
Formula Weight 450.15
Percent Purity 98%
Chemical Name or Material (2-Bromoethyl)triphenylphosphonium bromide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning