Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromoimidazole-4,5-dicarbonitrile, 97%

Click to view available options
Quantity:
250mg
1g
50847-09-1
C5HBrN4
196.995
MFCD00174288
WMPWSWMSTJAAPF-UHFFFAOYSA-N
2-bromo-4,5-dicyanoimidazole,2-bromoimidazole-4,5-dicarbonitrile,1h-imidazole-4,5-dicarbonitrile, 2-bromo,1h-imidazole-4,5-dicarbonitrile,2-bromo,2-bromo-1h-imidazole-4,5-dicarboxylic acid; dinitrile
2779148
2-bromo-1H-imidazole-4,5-dicarbonitrile
C(#N)C1=C(N=C(N1)Br)C#N
50847-09-1
C5HBrN4
196.995
MFCD00174288
WMPWSWMSTJAAPF-UHFFFAOYSA-N
2-bromo-4,5-dicyanoimidazole,2-bromoimidazole-4,5-dicarbonitrile,1h-imidazole-4,5-dicarbonitrile, 2-bromo,1h-imidazole-4,5-dicarbonitrile,2-bromo,2-bromo-1h-imidazole-4,5-dicarboxylic acid; dinitrile
2779148
2-bromo-1H-imidazole-4,5-dicarbonitrile
C(#N)C1=C(N=C(N1)Br)C#N
Color White
Quantity 250mg
UN Number UN3439
Beilstein 777309
Formula Weight 196.01
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Bromoimidazole-4,5-dicarbonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.