Alfa Aesar™ 2-Bromoimidazole, 95%

Manufacturer: Alfa Aesar™ H5255303

Quantity 1g

 View more versions of this product

Catalog No. AAH5255303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16681-56-4
    Molecular Formula C3H3BrN2
    Formula Weight 146.98
    MDL Number MFCD02179526
    Chemical Name or Material 2-Bromoimidazole

    Specifications

    Formula Weight 146.98
    Quantity 1g
    Molecular Formula C3H3BrN2
    MDL Number MFCD02179526
    Health Hazard 2 GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Melting Point 197°C
    CAS 16681-56-4
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material 2-Bromoimidazole
    Assay 95%

    Description

    Applications
    2-Bromoimidazole is used as an organic chemical synthesis intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H318-H302-H335-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit