Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromoindene, 98%

Click to view available options
Quantity:
1g
5g
25g
10485-09-3
C9H7Br
195.059
MFCD06797863
CCUYEVNCRQDQRF-UHFFFAOYSA-N
2-bromoindene,1h-indene, 2-bromo,indene, 2-bromo,2-bromanyl-1h-indene,2-bromoindene, 95+%,2-bromo-indene,pubchem9657,#,acmc-1c5rn,sodium perborate,tetrahydrate
575586
2-bromo-1H-indene
C1C2=CC=CC=C2C=C1Br
10485-09-3
C9H7Br
195.059
MFCD06797863
CCUYEVNCRQDQRF-UHFFFAOYSA-N
2-bromoindene,1h-indene, 2-bromo,indene, 2-bromo,2-bromanyl-1h-indene,2-bromoindene, 95+%,2-bromo-indene,pubchem9657,#,acmc-1c5rn,sodium perborate,tetrahydrate
575586
2-bromo-1H-indene
C1C2=CC=CC=C2C=C1Br
Applications
2-Bromoindene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point 35°C to 37°C
Quantity 1g
Formula Weight 195.06
Percent Purity 98%
Chemical Name or Material 2-Bromoindene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning