Alfa Aesar™ 2-Bromomesitylene, 99%

Manufacturer: Alfa Aesar™ A1227736

Quantity 500g

 View more versions of this product

Catalog No. AAA1227736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 576-83-0
    Molecular Formula C9H11Br
    Formula Weight 199.10g/mol
    MDL Number MFCD00000073
    Beilstein 1907245
    Synonym 2-Bromo-1,3,5-trimethylbenzene; 2,4,6-Trimethylbromobenzene
    Chemical Name or Material 2-Bromomesitylene

    Specifications

    Melting Point 2°C
    Boiling Point 224° to 226°C
    Quantity 500g
    Flash Point 96°C (204°F)
    CAS 576-83-0
    Molecular Formula C9H11Br
    Formula Weight 199.10g/mol
    MDL Number MFCD00000073
    Beilstein 1907245
    Synonym 2-Bromo-1,3,5-trimethylbenzene; 2,4,6-Trimethylbromobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Chemical Name or Material 2-Bromomesitylene
    Assay 99%

    Description

    Applications
    2-Bromomesitylene is a useful synthetic intermediate, Aromatics, Boron Derivatives, Catalyst. It is an intermediate used to prepare Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate (H714510) which is a frustrated phosphonium borate for metal-free catalytic hydrogenation of imines.

    Solubility
    Insoluble in water(20°C) .

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents, excess heat.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit