Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromomethyl-1,3-dioxolane, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
4360-63-8
C4H7BrO2
167
MFCD00003214
CKIIJIDEWWXQEA-UHFFFAOYSA-N
2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
78068
2-(bromomethyl)-1,3-dioxolane
C1COC(O1)CBr
4360-63-8
C4H7BrO2
167
MFCD00003214
CKIIJIDEWWXQEA-UHFFFAOYSA-N
2-bromomethyl-1,3-dioxolane, bromoacetaldehyde ethylene acetal, 1,3-dioxolane, 2-bromomethyl, 1,3-dioxolan-2-yl methyl bromide, bromomethyl dioxolane, acmc-1afrf, 2-bromomethyl-1,3-dioxolan, ksc495q7b, 2-bromomethyl-1,3 dioxolane, 2-bromomethyl-1, 3-dioxolane
78068
2-(bromomethyl)-1,3-dioxolane
C1COC(O1)CBr
Flash Point 62°C
Density 1.6100g/mL
Specific Gravity 1.61
Boiling Point 80°C to 82°C (27.0mmHg)
Refractive Index 1.4810 to 1.483
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Beilstein 19, II, 8
Formula Weight 167
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-1, 3-dioxolane, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-443-6

TSCA : TSCA