Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromomethyl-1,4-benzodioxane, 97%

Click to view available options
Quantity:
1g
5g
25g
2164-34-3
C9H9BrO2
229.073
MFCD00044831
QYLFKNVZIFTCIY-UHFFFAOYSA-N
2-bromomethyl-1,4-benzodioxane,2-bromomethyl-2,3-dihydro-1,4-benzodioxine,2-bromomethyl-2,3-dihydrobenzo b 1,4 dioxine,2-bromomethyl-2,3-dihydro-benzo 1,4 dioxine,2-bromomethyl-2,3-dihydrobenzo 1,4 dioxine,2-bromomethyl-2,3-dihydro-1,4-benzodioxin,3-bromomethyl-2,3-dihydro-1,4-benzodioxine,acmc-1cuiw,2-bromomethylbenzo-1,4-dioxan
98333
3-(bromomethyl)-2,3-dihydro-1,4-benzodioxine
C1C(OC2=CC=CC=C2O1)CBr
2164-34-3
C9H9BrO2
229.073
MFCD00044831
QYLFKNVZIFTCIY-UHFFFAOYSA-N
2-bromomethyl-1,4-benzodioxane,2-bromomethyl-2,3-dihydro-1,4-benzodioxine,2-bromomethyl-2,3-dihydrobenzo b 1,4 dioxine,2-bromomethyl-2,3-dihydro-benzo 1,4 dioxine,2-bromomethyl-2,3-dihydrobenzo 1,4 dioxine,2-bromomethyl-2,3-dihydro-1,4-benzodioxin,3-bromomethyl-2,3-dihydro-1,4-benzodioxine,acmc-1cuiw,2-bromomethylbenzo-1,4-dioxan
98333
3-(bromomethyl)-2,3-dihydro-1,4-benzodioxine
C1C(OC2=CC=CC=C2O1)CBr
Boiling Point 143°C to 146°C (8mmHg)
Quantity 25g
Beilstein 150154
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material 2-Bromomethyl-1,4-benzodioxane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning