Alfa Aesar™ 2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester, 95%

Manufacturer: Alfa Aesar™ H53309MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH53309MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 441011-76-3
    Molecular Formula C12H16BBrO2
    Formula Weight 282.97
    MDL Number MFCD04039022
    Synonym 2-(Bromomethyl)phenylboronic acid neopentyl glycol ester; 2-(Bromomethyl)phenylboronic acid mono(2,2-dimethylpropyl) ester
    Chemical Name or Material 2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester

    Specifications

    Formula Weight 282.97
    MDL Number MFCD04039022
    Synonym 2-(Bromomethyl)phenylboronic acid neopentyl glycol ester; 2-(Bromomethyl)phenylboronic acid mono(2,2-dimethylpropyl) ester
    Quantity 250mg
    Molecular Formula C12H16BBrO2
    CAS 441011-76-3
    Chemical Name or Material 2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit