Alfa Aesar™ 2-(Bromomethyl)benzoic acid, 97%

Manufacturer: Alfa Aesar™ H2892709

Quantity 10g

 View more versions of this product

Catalog No. AAH2892709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7115-89-1
    Molecular Formula C8H7BrO2
    Formula Weight 215.05
    MDL Number MFCD00090420
    UN Number UN3261
    Synonym alpha-Bromo-o-toluic acid; 2-Carboxybenzyl bromide
    Chemical Name or Material 2-(Bromomethyl)benzoic acid

    Specifications

    Formula Weight 215.05
    Molecular Formula C8H7BrO2
    MDL Number MFCD00090420
    UN Number UN3261
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    CAS 7115-89-1
    Melting Point ≈145°C
    Synonym alpha-Bromo-o-toluic acid; 2-Carboxybenzyl bromide
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2-(Bromomethyl)benzoic acid
    Assay 97%

    Description

    Applications
    2-(Bromomethyl)benzoic acid contains bi functional groups such as benzylic bromide and carboxylic acid are involved in nucleophilic reactions and esterification reactions respectively.

    Solubility
    Insoluble in water.

    Notes
    Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit