Alfa Aesar™ 2-(Bromomethyl)benzonitrile, 98%

Manufacturer: Alfa Aesar™ B2427822

Quantity 100g

 View more versions of this product

Catalog No. AAB2427822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22115-41-9
    Molecular Formula C8H6BrN
    Formula Weight 196.05
    MDL Number MFCD00001794
    UN Number UN2923
    Beilstein 742605
    Synonym alpha-Bromo-o-tolunitrile; 2-Cyanobenzyl bromide
    Chemical Name or Material 2-(Bromomethyl)benzonitrile

    Specifications

    Formula Weight 196.05
    MDL Number MFCD00001794
    UN Number UN2923
    Melting Point 70° to 74°C
    Synonym alpha-Bromo-o-tolunitrile; 2-Cyanobenzyl bromide
    Molecular Formula C8H6BrN
    CAS 22115-41-9
    Beilstein 742605
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material 2-(Bromomethyl)benzonitrile
    Assay 98%

    GHS H Statement
    H314-H318-H302-H312-H332-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit