Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Bromomethyl)phenylboronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
91983-14-1
C7H8BBrO2
214.85
MYVJCOQGXCONPE-UHFFFAOYSA-N
2-bromomethylphenylboronic acid,2-bromomethyl benzeneboronic acid,2-bromomethyl phenylboronic acid,2-bromomethyl phenyl boronic acid,2-bromomethylphenylboronicacid,2-boronobenzyl bromide,2-bromomethylphenyl boronic acid,o-boronobenzyl bromide,pubchem7781
2773278
91983-14-1
C7H8BBrO2
214.85
MYVJCOQGXCONPE-UHFFFAOYSA-N
2-bromomethylphenylboronic acid,2-bromomethyl benzeneboronic acid,2-bromomethyl phenylboronic acid,2-bromomethyl phenyl boronic acid,2-bromomethylphenylboronicacid,2-boronobenzyl bromide,2-bromomethylphenyl boronic acid,o-boronobenzyl bromide,pubchem7781
2773278
Melting Point 135°C to 136.5°C
Quantity 25g
Assay Percent Range 94% min. (HPLC)
Formula Weight 214.85
Percent Purity 95%
Chemical Name or Material 2-(Bromomethyl)phenylboronic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning