2-Bromopentane, tech. 90%

Manufacturer: Alfa Aesar A1720514

Quantity 25g

 View more versions of this product

Catalog No. AAA1720514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-81-3
    Molecular Formula C5H11Br
    Formula Weight 151.05
    MDL Number MFCD00000160
    UN Number UN2343
    Beilstein 1718829
    Chemical Name or Material 2-Bromopentane
    Grade Technical Grade

    Specifications

    Formula Weight 151.05
    Quantity 25g
    TSCA TSCA
    Boiling Point 117° to 118°C
    Melting Point −96°C
    Flash Point 20°C (68°F)
    Odor Characteristic
    Molecular Formula C5H11Br
    MDL Number MFCD00000160
    CAS 107-81-3
    UN Number UN2343
    Beilstein 1718829
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P280g-P273-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOPENTANE
    RTECS Number RZ9800000
    Chemical Name or Material 2-Bromopentane
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    Used as a chemical intermediate.

    Solubility
    Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol

    Notes
    Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P280g-P273-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit