Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopentane, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7023541
Click to view available options
Quantity:
100 g
500 g
107-81-3
C5H11Br
151.047
MFCD00000160
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
7890
2-bromopentane
CCCC(C)Br
107-81-3
C5H11Br
151.047
MFCD00000160
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
7890
2-bromopentane
CCCC(C)Br

Used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chemical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point -96°C
Density 1.22
Boiling Point 117°C to 118°C
Flash Point 20°C (68°F)
Odor Characteristic
Refractive Index 1.4403
Quantity 100 g
UN Number UN2343
Beilstein 1718829
Solubility Information Not miscible or difficult to mix in water. Soluble in Dichloromethane,Ether,Ethyl Acetate,Hexane and Methanol
Formula Weight 151.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Bromopentane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOPENTANE
EINECSNumber 203-521-3
RTECSNumber RZ9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit