Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenyl acetate, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
1829-37-4
C8H7BrO2
215.05
BEHBHYYPTOHUHX-UHFFFAOYSA-N
o-bromophenyl acetate,2-bromophenol acetate,1-acetoxy-2-bromobenzene,2-bromophenyl acetate,o-acetoxybromobenzene,phenol, 2-bromo-, acetate,phenol, o-bromo-, acetate,pubchem3055,2-bromo-phenol acetate
74586
(2-bromophenyl) acetate
CC(=O)OC1=CC=CC=C1Br
1829-37-4
C8H7BrO2
215.05
BEHBHYYPTOHUHX-UHFFFAOYSA-N
o-bromophenyl acetate,2-bromophenol acetate,1-acetoxy-2-bromobenzene,2-bromophenyl acetate,o-acetoxybromobenzene,phenol, 2-bromo-, acetate,phenol, o-bromo-, acetate,pubchem3055,2-bromo-phenol acetate
74586
(2-bromophenyl) acetate
CC(=O)OC1=CC=CC=C1Br
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 97.5 % min. (GC)
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Bromophenyl acetate

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS Signal Word: Warning

EINECSNumber : 217-380-