Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromophenylhydrazine hydrochloride, 95%

Click to view available options
Quantity:
5g
25g
100g
50709-33-6
C6H8BrClN2
223.498
MFCD00012926
PHCYUJRYSFMJMG-UHFFFAOYSA-N
2-bromophenylhydrazine hydrochloride,2-bromophenyl hydrazine hydrochloride,2-bromophenylhydrazine hcl,hydrazine, 2-bromophenyl-, monohydrochloride,1-2-bromophenyl hydrazine hydrochloride,1-bromo-2-hydrazino benzene hydrochloride,pubchem4463,acmc-1b1zz,ksc270g5h,2-bromophenylhydrazine, chloride
2723912
(2-bromophenyl)hydrazine;hydrochloride
C1=CC=C(C(=C1)NN)Br.Cl
50709-33-6
C6H8BrClN2
223.498
MFCD00012926
PHCYUJRYSFMJMG-UHFFFAOYSA-N
2-bromophenylhydrazine hydrochloride,2-bromophenyl hydrazine hydrochloride,2-bromophenylhydrazine hcl,hydrazine, 2-bromophenyl-, monohydrochloride,1-2-bromophenyl hydrazine hydrochloride,1-bromo-2-hydrazino benzene hydrochloride,pubchem4463,acmc-1b1zz,ksc270g5h,2-bromophenylhydrazine, chloride
2723912
(2-bromophenyl)hydrazine;hydrochloride
C1=CC=C(C(=C1)NN)Br.Cl
Quantity 5g
Melting Point ~188°C (decomposition)
Formula Weight 223.51
Percent Purity 95%
Chemical Name or Material 2-Bromophenylhydrazine hydrochloride

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger