Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromopropene, 99%, stab.

Click to view available options
Quantity:
5g
25g
100g
557-93-7
C3H5Br
120.977
MFCD00000140
PHMRPWPDDRGGGF-UHFFFAOYSA-N
2-bromopropene,isopropenyl bromide,1-propene, 2-bromo,2-bromo-1-propene,2-bromopropylene,propene, 2-bromo,isopropylene bromide,alpha-methylvinyl bromide,.alpha.-methylvinyl bromide,2-bromo-propene
11202
2-bromoprop-1-ene
CC(=C)Br
557-93-7
C3H5Br
120.977
MFCD00000140
PHMRPWPDDRGGGF-UHFFFAOYSA-N
2-bromopropene,isopropenyl bromide,1-propene, 2-bromo,2-bromo-1-propene,2-bromopropylene,propene, 2-bromo,isopropylene bromide,alpha-methylvinyl bromide,.alpha.-methylvinyl bromide,2-bromo-propene
11202
2-bromoprop-1-ene
CC(=C)Br
Applications
2-Bromopropene is used as a solvent and as an intermediate for organic synthesis. It is also used as a fumigant. Further, it reacts with nonanal to prepare 2-methyl-undec-1-en-3-ol using chromium(II) chloride as a reagent. It is utilized as a refrigerant used in organic synthesis.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
Refractive Index 1.443
Density 1.362
Sensitivity Light sensitive
Quantity 5g
Melting Point -87°C
Flash Point 4°C (39°F)
Odor Pungent
Boiling Point 47°C to 49°C
UN Number UN1993
Beilstein 1731926
Solubility Information Immiscible with water.
Formula Weight 120.98
Percent Purity 99%
Chemical Name or Material 2-Bromopropene, stabilized
Show More Show Less

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 209-185-4

RTECSNumber : UC7085000

TSCA : Yes

Recommended Storage : Keep cold