Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopropionitrile, 97%, Acros Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
19481-82-4
C3H4BrN
133.976
PYNYHMRMZOGVML-UHFFFAOYSA-N
2-bromopropionitrile,propanenitrile, 2-bromo,2-bromopropanitrile,propanenitrile, bromo,2-bromoproprionitrile,2-bromo-propionitrile,acmc-1bo3l,-2-bromopropionitrile,+ 2-bromo-propionitrile
140538
2-bromopropanenitrile
CC(C#N)Br
19481-82-4
C3H4BrN
133.976
PYNYHMRMZOGVML-UHFFFAOYSA-N
2-bromopropionitrile,propanenitrile, 2-bromo,2-bromopropanitrile,propanenitrile, bromo,2-bromoproprionitrile,2-bromo-propionitrile,acmc-1bo3l,-2-bromopropionitrile,+ 2-bromo-propionitrile
140538
2-bromopropanenitrile
CC(C#N)Br
Flash Point 58°C
Packaging Glass bottle
Quantity 5g
Boiling Point 68.0°C to 69.0°C (67.0hPa)
Density 1.5500g/mL
Linear Formula CH3CH(Br)CN
Formula Weight 133.97
Chemical Name or Material 2-Bromopropionitrile