Learn More
2-Bromopropionyl bromide, 97%, Thermo Scientific Chemicals
Description
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.
Solubility
Miscible with trichloromethane, diethyl ether, benzene and acetone.
Notes
Incompatible with water, alcohols, oxidizing agents and strong alkalis.
Specifications
Density | 2.061 |
Boiling Point | 48°C to 50°C (10 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.521 |
Quantity | 500 g |
UN Number | UN3265 |
Beilstein | 1071331 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with trichloromethane,diethyl ether,benzene and acetone. |
Formula Weight | 215.88 |
Show More |
Safety and Handling
Hazard Category | H302-H314-H317-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 209-261-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.