Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromopropionyl chloride, 98%

Click to view available options
Quantity:
10g
50g
250g
7148-74-5
C3H4BrClO
171.418
MFCD00000711
OZGMODDEIHYPRY-UHFFFAOYSA-N
2-bromopropionyl chloride,propanoyl chloride, 2-bromo,2-bromopropionylchloride,2-bromo-propionyl chloride,2-brominepropionylchloride,alpha-bromopropionyl chloride,2-bromopropionic acid chloride,sgqdjlpqbsmuh@,2-bromopropanoylchloride,acmc-1cjhb
97980
2-bromopropanoyl chloride
CC(C(=O)Cl)Br
7148-74-5
C3H4BrClO
171.418
MFCD00000711
OZGMODDEIHYPRY-UHFFFAOYSA-N
2-bromopropionyl chloride,propanoyl chloride, 2-bromo,2-bromopropionylchloride,2-bromo-propionyl chloride,2-brominepropionylchloride,alpha-bromopropionyl chloride,2-bromopropionic acid chloride,sgqdjlpqbsmuh@,2-bromopropanoylchloride,acmc-1cjhb
97980
2-bromopropanoyl chloride
CC(C(=O)Cl)Br
Applications
2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.

Solubility
Reacts with water.

Notes
Protect from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep away from ignition sources. Protect from electrostatic charges.
Boiling Point 131°C to 133°C
Quantity 250g
Flash Point 51°C (125°F)
UN Number UN2920
Beilstein 773800
Formula Weight 171.43
Percent Purity 98%
Chemical Name or Material 2-Bromopropionyl chloride

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.