Learn More
2-Bromopropionyl chloride, 98%, Thermo Scientific Chemicals
Description
2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromopropionyl chloride is a reagent that is used in the preparation of pyrazinecarboxamide-based inhibitors of diacylglycerol acyltransferase. It is also used in the synthesis of 2-aminopropionyl polystyrene resin by Friedel Crafts reaction.
Solubility
Reacts with water.
Notes
Protect from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep away from ignition sources. Protect from electrostatic charges.
Specifications
Density | 1.7 |
Boiling Point | 131°C to 133°C |
Flash Point | 51°C (125°F) |
Refractive Index | 1.478 |
Quantity | 10 g |
UN Number | UN2920 |
Beilstein | 773800 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 171.43 |
Show More |
Safety and Handling
Hazard Category | H226-H314 |
Hazard Statement | GHS H Statement H314-H318-H226 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
EINECSNumber | 230-466-2 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.