Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromopyridine-3-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
35905-85-2
C6H4BrNO2
202.01
MFCD01646068
MMWNKXIFVYQOTK-UHFFFAOYSA-N
2-bromonicotinic acid,2-bromo-3-pyridinecarboxylic acid,3-pyridinecarboxylic acid, 2-bromo,2-bromo-nicotinic acid,2-bromo-3-carboxypyridine,3-bromopyridine-3-carboxylic acid,bromo-nicotinic acid,pubchem5071,acmc-1aj4p,ksc223s8f
233262
2-bromopyridine-3-carboxylic acid
C1=CC(=C(N=C1)Br)C(=O)O
35905-85-2
C6H4BrNO2
202.01
MFCD01646068
MMWNKXIFVYQOTK-UHFFFAOYSA-N
2-bromonicotinic acid,2-bromo-3-pyridinecarboxylic acid,3-pyridinecarboxylic acid, 2-bromo,2-bromo-nicotinic acid,2-bromo-3-carboxypyridine,3-bromopyridine-3-carboxylic acid,bromo-nicotinic acid,pubchem5071,acmc-1aj4p,ksc223s8f
233262
2-bromopyridine-3-carboxylic acid
C1=CC(=C(N=C1)Br)C(=O)O
Quantity 5g
Packaging Glass bottle
Melting Point 200.0°C to 203.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 202.01
Percent Purity 97%
Chemical Name or Material 2-Bromopyridine-3-carboxylic acid

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning