Alfa Aesar™ 2-Bromostyrene, 97%, stab. with ca 0.05% 4-tert-butylcatechol

Manufacturer: Alfa Aesar™ B2418114

Quantity 25g

 View more versions of this product

Catalog No. AAB2418114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2039-88-5
    Molecular Formula C8H7Br
    Formula Weight 183.05
    MDL Number MFCD00000076
    Beilstein 2323537
    Chemical Name or Material 2-Bromostyrene, stabilized with ≈0.05% 4-tert-butylcatechol

    Specifications

    Formula Weight 183.05
    Molecular Formula C8H7Br
    MDL Number MFCD00000076
    CAS 2039-88-5
    Melting Point −53°C
    Beilstein 2323537
    Boiling Point 102° to 104°C (22mmHg)
    Quantity 25g
    Flash Point 85°C (185°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material 2-Bromostyrene, stabilized with ≈0.05% 4-tert-butylcatechol
    Assay 97%

    Description

    Applications
    2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.

    Solubility
    Miscible with methanol.

    Notes
    Incompatible with acids, bases, oxidizing agents and halogens.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit