Learn More
2-Bromostyrene, 96%, stab. with ca 0.05% 4-tert-butylcatechol, Thermo Scientific Chemicals
Description
2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromostyrene is used as a cross linking agent in fire-resistant bromostyrene-crosslinked polyesters. Further, it is used to prepare trans-1-(2-bromophenyl)-2-(3-bromophenyl)ethene by reacting with 1-bromo-3-iodo-benzene, involving palladium(II) acetate as catalyst.
Solubility
Miscible with methanol.
Notes
Incompatible with acids, bases, oxidizing agents and halogens.
Specifications
Melting Point | -53°C |
Density | 1.416 |
Boiling Point | 102°C to 104°C (22 mmHg) |
Flash Point | 85°C (185°F) |
Refractive Index | 1.592 |
Quantity | 5 g |
Beilstein | 2323537 |
Solubility Information | Miscible with methanol. |
Formula Weight | 183.05 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 218-027-3 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.