Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoterephthalic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
586-35-6
C8H5BrO4
245.03
MFCD00002403
QPBGNSFASPVGTP-UHFFFAOYSA-N
bromoterephthalic acid,2-bromobenzene-1,4-dicarboxylic acid,2-bromoterephthalicacid,2-bromo-terephthalic acid,2-bromo-1,4-benzenedicarboxylic acid,1,4-benzenedicarboxylic acid, 2-bromo,labotest-bb lt00454996,pubchem12798,2-bromo terephthalic acid,acmc-1aoy9
68513
2-bromoterephthalic acid
C1=CC(=C(C=C1C(=O)O)Br)C(=O)O
586-35-6
C8H5BrO4
245.03
MFCD00002403
QPBGNSFASPVGTP-UHFFFAOYSA-N
bromoterephthalic acid,2-bromobenzene-1,4-dicarboxylic acid,2-bromoterephthalicacid,2-bromo-terephthalic acid,2-bromo-1,4-benzenedicarboxylic acid,1,4-benzenedicarboxylic acid, 2-bromo,labotest-bb lt00454996,pubchem12798,2-bromo terephthalic acid,acmc-1aoy9
68513
2-bromoterephthalic acid
C1=CC(=C(C=C1C(=O)O)Br)C(=O)O
Packaging Glass bottle
Melting Point 295.0°C to 297.0°C
Quantity 25g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula BrC6H3(CO2H)2
Beilstein 09,848
Formula Weight 245.03
Percent Purity 95%
Chemical Name or Material 2-Bromoterephthalic acid, 95%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-572-8