Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromothiazole-5-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
54045-76-0
C4H2BrNO2S
208.03
MFCD04115730
BESGTWHUMYHYEQ-UHFFFAOYSA-N
2-bromothiazole-5-carboxylic acid,2-bromo-5-thiazolecarboxylic acid,2-bromo-5-thiazolecarboxylicacid,2-bromo-thiazole-5-carboxylic acid,2-bromo-5-carboxythiazole,5-thiazolecarboxylic acid, 2-bromo,2-bromthiazol-5-carboxylic acid,pubchem15963,acmc-1ap3n,2-bromothiazole-5-carboxylic
2763210
2-bromo-1,3-thiazole-5-carboxylic acid
C1=C(SC(=N1)Br)C(=O)O
54045-76-0
C4H2BrNO2S
208.03
MFCD04115730
BESGTWHUMYHYEQ-UHFFFAOYSA-N
2-bromothiazole-5-carboxylic acid,2-bromo-5-thiazolecarboxylic acid,2-bromo-5-thiazolecarboxylicacid,2-bromo-thiazole-5-carboxylic acid,2-bromo-5-carboxythiazole,5-thiazolecarboxylic acid, 2-bromo,2-bromthiazol-5-carboxylic acid,pubchem15963,acmc-1ap3n,2-bromothiazole-5-carboxylic
2763210
2-bromo-1,3-thiazole-5-carboxylic acid
C1=C(SC(=N1)Br)C(=O)O
Quantity 1g
Color White
Melting Point 174.5°C to 175.5°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 208.03
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Bromothiazole-5-carboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning