Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Bromothien-3-yl)methanol, 97%, Thermo Scientific™

Catalog Number cc56609da
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
70260-16-1
C5H5BrOS
193.058
MFCD09025881
BOJYRNFSHPAZBL-UHFFFAOYSA-N
2-bromothiophen-3-yl methanol, 2-bromo-3-hydroxymethyl-thiophene, 2-bromothien-3-yl methanol, 2-bromo-3-thiophenemethanol, pubchem22131, 2-bromo-3-thienyl methanol, 3-thiophenemethanol, 2-bromo, 2-bromo-3-hydroxymethylthiophene, 2-bromo-3-thienyl methan-1-ol, 2-bromo-thiophen-3-yl-methanol
12479010
(2-bromothiophen-3-yl)methanol
C1=CSC(=C1CO)Br
70260-16-1
C5H5BrOS
193.058
MFCD09025881
BOJYRNFSHPAZBL-UHFFFAOYSA-N
2-bromothiophen-3-yl methanol, 2-bromo-3-hydroxymethyl-thiophene, 2-bromothien-3-yl methanol, 2-bromo-3-thiophenemethanol, pubchem22131, 2-bromo-3-thienyl methanol, 3-thiophenemethanol, 2-bromo, 2-bromo-3-hydroxymethylthiophene, 2-bromo-3-thienyl methan-1-ol, 2-bromo-thiophen-3-yl-methanol
12479010
(2-bromothiophen-3-yl)methanol
C1=CSC(=C1CO)Br
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.06
Percent Purity 95%
Chemical Name or Material (2-Bromothien-3-yl)methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit