Learn More
2-Bromothiophene, 98+%, Thermo Scientific Chemicals
Description
2-Bromothiophene has been used in electrochemical reduction of a number of mono- and dihalothiophenes at carbon cathodes in dimethylformamide containing tetramethylammonium per chlorate by cyclic voltammetry and controlled-potential electrolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromothiophene has been used in electrochemical reduction of a number of mono- and dihalothiophenes at carbon cathodes in dimethylformamide containing tetramethylammonium per chlorate by cyclic voltammetry and controlled-potential electrolysis.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
Specifications
Melting Point | <-10°C |
Density | 1.701 |
Boiling Point | 149°C to 152°C |
Flash Point | 58°C (136°F) |
Refractive Index | 1.586 |
Quantity | 50 g |
UN Number | UN2929 |
Beilstein | 104663 |
Sensitivity | Light sensitive |
Solubility Information | Not miscible or difficult to mix in water. |
Show More |
Safety and Handling
Hazard Category | H226-H300+H310+H330-H315-H318 |
Hazard Statement | GHS H Statement H300-H310-H330-H226-H315-H319 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S. |
EINECSNumber | 213-699-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.