Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromothiophenol, 97%

Click to view available options
Quantity:
5g
25g
6320-02-1
C6H5BrS
189.07
MFCD00004827
YUQUNWNSQDULTI-UHFFFAOYSA-N
2-bromothiophenol,o-bromothiophenol,o-bromobenzenethiol,2-bromo thiophenol,benzenethiol, 2-bromo,benzenethiol, o-bromo,2-bromobenzene-1-thiol,2-bromo-benzenethiol,2-bromo-thiophenol,2-bromobenzene thiol
80598
2-bromobenzenethiol
C1=CC=C(C(=C1)S)Br
6320-02-1
C6H5BrS
189.07
MFCD00004827
YUQUNWNSQDULTI-UHFFFAOYSA-N
2-bromothiophenol,o-bromothiophenol,o-bromobenzenethiol,2-bromo thiophenol,benzenethiol, 2-bromo,benzenethiol, o-bromo,2-bromobenzene-1-thiol,2-bromo-benzenethiol,2-bromo-thiophenol,2-bromobenzene thiol
80598
2-bromobenzenethiol
C1=CC=C(C(=C1)S)Br
Refractive Index 1.635
Sensitivity Air Sensitive
Density 1.604
Boiling Point 114°C to 116°C (10mmHg)
Flash Point >110°C (230°F)
Quantity 25g
Odor Unpleasant
UN Number UN2922
Beilstein 2039769
Formula Weight 189.08
Percent Purity 97%
Chemical Name or Material 2-Bromothiophenol
Show More Show Less

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 228-665-4

TSCA : Yes

Recommended Storage : Ambient temperatures