Alfa Aesar™ 2-Bromotoluene, 99%

Manufacturer: Alfa Aesar™ A1231536

Quantity 500g

 View more versions of this product

Catalog No. AAA1231536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95-46-5
    Molecular Formula C7H7Br
    Formula Weight 171.04
    MDL Number MFCD00000068
    Beilstein 1904176
    Merck Index 14,1439
    Synonym 2-Methylbromobenzene
    Chemical Name or Material 2-Bromotoluene

    Specifications

    Formula Weight 171.04
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1904176
    Melting Point −27°C
    Boiling Point 179° to 181°C
    Quantity 500g
    Flash Point 78°C (172°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    RTECS Number XS7965500
    TSCA TSCA
    Molecular Formula C7H7Br
    CAS 95-46-5
    Merck Index 14,1439
    MDL Number MFCD00000068
    Synonym 2-Methylbromobenzene
    Chemical Name or Material 2-Bromotoluene
    Assay 99%

    Description

    Applications
    2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

    Solubility
    Soluble in alcohol, ether, acetone, and ethanol. Insoluble in water.

    Notes
    Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.

    GHS H Statement
    H302-H315-H319-H335-H227
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit