Learn More
(+/-)-2-Butanol, 99%, Thermo Scientific Chemicals
Description
(±)-2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(^+)-2-Butanol is widely used as a solvent for domestic cleaning agents and paint removers. It is also utilized in perfumes and artificial flavors. It plays an important role in the extraction of fish meal to produce fish protein concentrate. It is involved in the production of fruit essences. It plays an essential role in the esterification of acetic acid. It is useful for the derivatization of chlorinated hydroxyfuranones. It is also used as a precursor for methyl ethyl ketone.
Solubility
Miscible with methanol and diethyl ether. Slightly miscible with water.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -115°C |
Density | 0.808 |
Boiling Point | 98°C to 101°C |
Quantity | 500 mL |
Flash Point | 26°C (79°F) |
Odor | Sweet |
Refractive Index | 1.397 |
UN Number | UN1120 |
Beilstein | 1718765 |
Merck Index | 14,1541 |
Show More |
Safety and Handling
Hazard Category | H226-H319-H335-H336 |
Hazard Statement | GHS H Statement H226-H319-H335-H336 Flammable liquid and vapour. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS |
EINECSNumber | 201-158-5 |
RTECSNumber | EO1750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.