Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Riedel-de Haen™ 2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC), Honeywell™ Riedel-de Haen™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass Bottle
78-92-2
C4H10O
74.123
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYSA-N
2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol
6568
CHEBI:35687
butan-2-ol
CCC(C)O
78-92-2
C4H10O
74.123
MFCD00004569
BTANRVKWQNVYAZ-UHFFFAOYSA-N
2-butanol,sec-butanol,sec-butyl alcohol,2-hydroxybutane,2-butyl alcohol,s-butyl alcohol,butylene hydrate,s-butanol,1-methyl-1-propanol,1-methylpropyl alcohol
6568
CHEBI:35687
butan-2-ol
CCC(C)O

Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Linear Formula CH3CH2CH(OH)CH3
UN Number UN1120
Beilstein 773649
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Density 0.808g/mL at 25°C
Explosion Limit 0.098
Packaging Glass Bottle
Refractive Index n20/D 1.397
Vapor Pressure 12.5mmHg (20°C)
Vapor Density 2.6 (vs air)
Flash Point 27°C (80.6°F)
Physical State Liquid
Boiling Point 99°C
Color Colorless
Melting Point -115°C
Quantity 250mL
Chemical Name or Material 2-Butanol, Reag. Ph. Eur.
Show More Show Less

P210-P305 + P351 + P338-P370 + P378

H226-H319-H335-H336

DOTInformation : Class 3 - PG 3 - Butanols

EINECSNumber : 201-158-5

RTECSNumber : EO1750000