Alfa Aesar™ 2-Butanone, 97%

Manufacturer: Alfa Aesar™ 31069K7

Quantity 4L

 View more versions of this product

Catalog No. AA31069K7

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 78-93-3
    Linear Formula CH3COCH2CH3
    Formula Weight 72.11
    MDL Number MFCD00011648
    UN Number UN1193
    Beilstein 741880
    Merck Index 14,6072
    Synonym Methyl ethyl ketone; MEK
    Chemical Name or Material 2-Butanone

    Specifications

    Formula Weight 72.11
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Characteristic
    Health Hazard 2 GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    RTECS Number EL6475000
    Refractive Index 1.3790
    Boiling Point 80°C
    Merck Index 14,6072
    Health Hazard 1 Danger
    Density 0.805g/mL
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL METHYL KETONE
    Flash Point −3° (26°F)
    Color Colorless
    Melting Point −87°C
    Linear Formula CH3COCH2CH3
    UN Number UN1193
    Beilstein 741880
    Synonym Methyl ethyl ketone; MEK
    TSCA TSCA
    Physical Form Liquid
    Quantity 4L
    MDL Number MFCD00011648
    CAS 78-93-3
    Chemical Name or Material 2-Butanone
    Assay 97%

    GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit