Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, +99%, for spectroscopy, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass bottle
78-93-3
C4H8O
72.11
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(C)=O
78-93-3
C4H8O
72.11
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(C)=O
For UV/VIS spectroscopy
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 at 350nm, 0.1 at 340nm, 1 at 330nm
CAS Min % 99.0
Absorbance Conditions (1 cm cell vs water)
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C2H5COCH3
Beilstein 01, IV, 3243
Fieser 01,678
Merck Index 15, 6143
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Physical Form Liquid
Percent Purity ≥99%
Grade Spectroscopy
Density 0.8060g/mL
Packaging Glass bottle
Refractive Index 1.3780 to 1.3800
Residue after Evaporation 0.001% max.
Solubility Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Specific Gravity 0.806
Water 0.1% max.
Flash Point −7°C
Viscosity 0.42 mPa.s (15°C)
Boiling Point 80.0°C
Color Undesignated
Melting Point -87.0°C
Quantity 500mL
Chemical Name or Material 2-Butanone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-159-0

RTECSNumber : EL6475000

TSCA : TSCA