Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Riedel-de Haen™ 2-Butanone, puriss. p.a. ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haen™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass Bottle

Puriss. p.a., ACS reagent,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Appearance of Solution Colorless
Density 0.805g/mL at 25°C
Packaging Glass Bottle
Refractive Index n20/D 1.379(lit.)
Flash Point −3°C (26.6°F)
Boiling Point 79°C to 81°C
Melting Point -87°C
Quantity 1L