Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
78-93-3
C4H8O
72.11
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
CCC(C)=O
78-93-3
C4H8O
72.11
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
butan-2-one
CCC(C)=O

M1260, 78-93-3, C4H8O

Spectrum™ 2-Butanone, Reagent, ACS
CAS Max % 1
UN Number UN1193
Percent Purity 99%
Packaging Steel Can
Residue after Evaporation 2.5e-005
Titratable Acid 0.0005 meq/g
Water 0.002
Quantity 20 L