Alfa Aesar™ 2-Buten-1-ol, cis + trans (ca 1:19), 96%

Manufacturer: Alfa Aesar™ A1068122

Quantity 100g

 View more versions of this product

Catalog No. AAA1068122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6117-91-5
    Molecular Formula CH3CH=CHCH2OH
    Formula Weight 72.11
    MDL Number MFCD00002923
    UN Number UN1987
    Beilstein 1719374
    Merck Index 14,2601
    Synonym Crotyl alcohol; 1-Hydroxybut-2-ene
    Chemical Name or Material 2-Buten-1-ol, cis + trans (≈ 1:19)

    Specifications

    Formula Weight 72.11
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Warning
    Synonym Crotyl alcohol; 1-Hydroxybut-2-ene
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H226-H302-H312
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    MDL Number MFCD00002923
    UN Number UN1987
    Beilstein 1719374
    Boiling Point 121° to 122°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
    Molecular Formula CH3CH=CHCH2OH
    CAS 6117-91-5
    Merck Index 14,2601
    Flash Point 37°C (98°F)
    RTECS Number EM9275000
    Chemical Name or Material 2-Buten-1-ol, cis + trans (≈ 1:19)
    Assay 96%

    Description

    Applications
    2-Buten-1-ol, cis + trans (ca 1:19) is used as a building block for the synthesis of various pharmaceutical compounds such as crotyl mercaptan.

    Solubility
    Miscible with water.

    Notes
    Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides, acid anhydrides and phosphorus halides.

    GHS H Statement
    H226-H302-H312
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit