Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Buten-1-ol, cis + trans (ca 1:19), 96%

Manufacturer:  Alfa Aesar™ A1068114

Quantity 25g

 View more versions of this product

Catalog No. AAA1068114

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
2-Buten-1-ol, cis + trans (ca 1:19) is used as a building block for the synthesis of various pharmaceutical compounds such as crotyl mercaptan.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides, acid anhydrides and phosphorus halides.
Description & Specifications

Chemical Identifiers

6117-91-5
72.11
UN1987
14,2601
2-Buten-1-ol, cis + trans (≈ 1:19)
CH3CH=CHCH2OH
MFCD00002923
1719374
Crotyl alcohol; 1-Hydroxybut-2-ene

Specifications

6117-91-5
72.11
UN1987
14,2601
121° to 122°C
37°C (98°F)
96%
CH3CH=CHCH2OH
MFCD00002923
1719374
Crotyl alcohol; 1-Hydroxybut-2-ene
25g
2-Buten-1-ol, cis + trans (≈ 1:19)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H302-H312
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.

RTECSNumber : EM9275000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit