Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-4-chloro-5-formylimidazole, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
83857-96-9
C8H11ClN2O
186.639
MFCD01934396
JLVIHQCWASNXCK-UHFFFAOYSA-N
2-butyl-4-chloro-5-formylimidazole,2-butyl-4-chloro-1h-imidazole-5-carbaldehyde,unii-7h170u0swe,2-butyl-5-chloroimidazole-4-carbaldehyde,2-butyl-5-chloro-3h-imidazole-4-carbaldehyde,2-butyl-4 5-chloro-5 4 imidazolecarboxaldehyde,1h-imidazole-4-carboxaldehyde, 2-butyl-5-chloro,2-butyl-4-chloro-1h-imidazole-5-carboxaldehyde,2-butyl-4-chloro-5-formylimidazloe,2-n-butyl-4-chloro-5-formylimidazole
55176
2-butyl-5-chloro-1H-imidazole-4-carbaldehyde
CCCCC1=NC(=C(N1)Cl)C=O
83857-96-9
C8H11ClN2O
186.639
MFCD01934396
JLVIHQCWASNXCK-UHFFFAOYSA-N
2-butyl-4-chloro-5-formylimidazole,2-butyl-4-chloro-1h-imidazole-5-carbaldehyde,unii-7h170u0swe,2-butyl-5-chloroimidazole-4-carbaldehyde,2-butyl-5-chloro-3h-imidazole-4-carbaldehyde,2-butyl-4 5-chloro-5 4 imidazolecarboxaldehyde,1h-imidazole-4-carboxaldehyde, 2-butyl-5-chloro,2-butyl-4-chloro-1h-imidazole-5-carboxaldehyde,2-butyl-4-chloro-5-formylimidazloe,2-n-butyl-4-chloro-5-formylimidazole
55176
2-butyl-5-chloro-1H-imidazole-4-carbaldehyde
CCCCC1=NC(=C(N1)Cl)C=O
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 96°C to 100°C
Quantity 25g
Loss on Drying 0.5% max.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 186.64
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Butyl-4-chloro-5-formylimidazole
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning