Alfa Aesar™ 2-Butynyl acetate, 98%

Manufacturer: Alfa Aesar™ B2458406

Quantity 5g

 View more versions of this product

Catalog No. AAB2458406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34485-37-5
    Molecular Formula CH3CO2CH2C≡CCH3
    Formula Weight 112.13
    MDL Number MFCD00078282
    UN Number UN3272
    Beilstein 1745269
    Synonym Acetic acid 2-butynyl ester; 1-Acetoxy-2-butyne
    Chemical Name or Material 2-Butynyl acetate

    Specifications

    Formula Weight 112.13
    Molecular Formula CH3CO2CH2C≡CCH3
    MDL Number MFCD00078282
    UN Number UN3272
    Beilstein 1745269
    Synonym Acetic acid 2-butynyl ester; 1-Acetoxy-2-butyne
    Health Hazard 1 Warning
    Boiling Point 150° to 152°C (680mmHg)
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Quantity 5g
    Flash Point 56°C (132°F)
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    CAS 34485-37-5
    Chemical Name or Material 2-Butynyl acetate
    Assay 98%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit