Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Carboxycinnamic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
612-40-8
C10H8O4
192.17
MFCD00004380
SCWPNMHQRGNQHH-AATRIKPKSA-N
2-carboxycinnamic acid,2-2-carboxyvinyl benzoic acid,2-2-carboxyethenyl benzoic acid,e-2-2-carboxyvinyl benzoic acid,2-e-3-hydroxy-3-oxoprop-1-enyl benzoic acid,2-1e-2-carboxyvinyl benzoic acid,2-e-2-carboxyethenyl benzoic acid,2-1e-2-carboxyeth-1-en-1-yl benzoic acid,2-e-2-carboxyvinyl benzoicacid,2-carboxy cinnamic acid
904938
2-[(E)-2-carboxyethenyl]benzoic acid
C1=CC=C(C(=C1)C=CC(=O)O)C(=O)O
612-40-8
C10H8O4
192.17
MFCD00004380
SCWPNMHQRGNQHH-AATRIKPKSA-N
2-carboxycinnamic acid,2-2-carboxyvinyl benzoic acid,2-2-carboxyethenyl benzoic acid,e-2-2-carboxyvinyl benzoic acid,2-e-3-hydroxy-3-oxoprop-1-enyl benzoic acid,2-1e-2-carboxyvinyl benzoic acid,2-e-2-carboxyethenyl benzoic acid,2-1e-2-carboxyeth-1-en-1-yl benzoic acid,2-e-2-carboxyvinyl benzoicacid,2-carboxy cinnamic acid
904938
2-[(E)-2-carboxyethenyl]benzoic acid
C1=CC=C(C(=C1)C=CC(=O)O)C(=O)O
Infrared Spectrum Authentic
Quantity 5g
Color White
Melting Point 197°C to 202°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 192.17
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Carboxycinnamic acid, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-308-9

TSCA : TSCA