Learn More
MP Biomedical™ 2-CAT Fast Track ELISA Kit
Quantitative Enzyme-linked Immunosorbent Assay
Manufacturer:  MP Biomedical™ 07LE6500
Catalog No. MP07LE6500
Add to cart
Description
Description
The 2-CAT Fast Track ELISA Kit is intended for the measurement of Adrenaline (epinephrine) and Noradrenaline (norepinephrine) in plasma and urine. Samples are extracted by using a cis-diol-specific affinity gel, acylated and then derivatized enzymatically.
Specifications
Specifications
Kit and Assay
FDA Exempt
2°C to 8°C
Human
Biogenic Amines
10 μL for Urin; 300 μL for Plasma
96 Wells
Plasma, Urine