Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
6609-64-9
C2H4ClO3P
142.48
MFCD00043138
SBMUNILHNJLMBF-UHFFFAOYSA-N
2-chloro-2-oxo-1,3,2-dioxaphospholane,ethylene chlorophosphate,2-chloro-1,3,2-dioxaphospholane-2-oxide,2-chloro-1,3,2-dioxaphospholane 2-oxide,ethylene glycol chlorophosphate,soqdebhb^bsvusup@,2-chloro-1,3,2??-dioxaphospholan-2-one,2-chlor-1,3,2-dioxaphospholan-2-oxid,acmc-1b2u0,ethylene cycl-chlorophosphate
81087
2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
C1COP(=O)(O1)Cl
6609-64-9
C2H4ClO3P
142.48
MFCD00043138
SBMUNILHNJLMBF-UHFFFAOYSA-N
2-chloro-2-oxo-1,3,2-dioxaphospholane,ethylene chlorophosphate,2-chloro-1,3,2-dioxaphospholane-2-oxide,2-chloro-1,3,2-dioxaphospholane 2-oxide,ethylene glycol chlorophosphate,soqdebhb^bsvusup@,2-chloro-1,3,2??-dioxaphospholan-2-one,2-chlor-1,3,2-dioxaphospholan-2-oxid,acmc-1b2u0,ethylene cycl-chlorophosphate
81087
2-chloro-1,3,2$l^{5}-dioxaphospholane 2-oxide
C1COP(=O)(O1)Cl
Flash Point >110°C
Density 1.5500g/mL
Boiling Point 98.0°C to 99.0°C (2.0mmHg)
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 11.0°C to 14.0°C
Color White
Refractive Index 1.4490 to 1.4540
Specific Gravity 1.55
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 142.48
Physical Form Solid or Film
Percent Purity 95%
Chemical Name or Material 2-Chloro-1, 3, 2-dioxaphospholane-2-oxide
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger