Alfa Aesar™ 2-Chloro-1,3-difluoro-4-nitrobenzene, 97%

Manufacturer: Alfa Aesar™ H3380614

Quantity 25g

 View more versions of this product

Catalog No. AAH3380614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3847-58-3
    Molecular Formula C6H2ClF2NO2
    Formula Weight 193.54
    MDL Number MFCD00042201
    UN Number UN2923
    Synonym 3-Chloro-2,4-difluoronitrobenzene
    Chemical Name or Material 2-Chloro-1,3-difluoro-4-nitrobenzene

    Specifications

    Formula Weight 193.54
    Molecular Formula C6H2ClF2NO2
    MDL Number MFCD00042201
    CAS 3847-58-3
    UN Number UN2923
    Synonym 3-Chloro-2,4-difluoronitrobenzene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Melting Point 45° to 49°C
    Quantity 25g
    Chemical Name or Material 2-Chloro-1,3-difluoro-4-nitrobenzene
    Assay 97%

    GHS H Statement
    H314-H318-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit