Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
153433-26-2
C5H10ClN2·BF4
220.41
CIB
153433-26-2
C5H10ClN2·BF4
220.41
CIB
Coupling reagent
Packaging Glass Bottle
Color White
Infrared Spectrum Authentic
Quantity 1g
Melting Point 58.0°C to 60.0°C
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Formula Weight 220.41
Physical Form Powder or Crystalline Powder
Percent Purity 96%
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolidinium tetrafluoroborate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning