Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1,3-dimethylimidazolinium chloride, 90%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
37091-73-9
C5H10Cl2N2
169.05
AEBBXVHGVADBHA-UHFFFAOYSA-M
2-chloro-1,3-dimethylimidazolinium chloride,unii-56ka2a1u1d,2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride,dmc mi,2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride,pubchem16443,2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride,1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1,2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
10176306
2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
CN1CC[N+](=C1Cl)C.[Cl-]
37091-73-9
C5H10Cl2N2
169.05
AEBBXVHGVADBHA-UHFFFAOYSA-M
2-chloro-1,3-dimethylimidazolinium chloride,unii-56ka2a1u1d,2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride,dmc mi,2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride,pubchem16443,2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride,1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1,2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
10176306
2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
CN1CC[N+](=C1Cl)C.[Cl-]
Coupling reagent
Melting Point 112°C to 121°C
Elemental Analysis 31.26 to 39.79% C, 14.58 to 18.56% N
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (HPLC)
Merck Index 15,3434
Formula Weight 169.05
Physical Form Powder or Crystalline Powder
Percent Purity 90%
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolinium chloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning