Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-1,3-dimethylimidazolium tetrafluoroborate, 98+%

Manufacturer:  Alfa Aesar™ H5431603

Quantity 1g

 View more versions of this product

Catalog No. AAH5431603


  Check Availability
Add to cart

Specifications

Description

Applications
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate is a reagent for esterification of C-terminal amino acids to Wang resin without racemization and for coupling α,α-dimethyl amino acids; and for preparation of the benzotriazol-1-yloxy derivative, a new coupling reagent. It is used as a reagent for esterification and peptide coupling of sterically hindered amino acids; preparation of efficient diazo-transfer reagents; oxidative insertion for palladium and nickel catalyst synthesis; and preparation of catalysts for use in coupling reactions.

Solubility
Soluble in acetonitrile (0.1 g/ml).

Notes
Moisture and light sensitive. Incompatible with oxidizing agents and heat.
Description & Specifications

Chemical Identifiers

153433-26-2
220.4
CIB
C5H10ClN2·BF4
MFCD09039291
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate

Specifications

175° to 177°C
153433-26-2
220.4
CIB
≥98%
1g
C5H10ClN2·BF4
MFCD09039291
2-Chloro-1,3-dimethylimidazolium tetrafluoroborate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit