Alfa Aesar™ 2-Chloro-1,4-di-n-propoxybenzene, 97%

Manufacturer: Alfa Aesar™ B2477609

Quantity 10g

 View more versions of this product

Catalog No. AAB2477609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 916791-98-5
    Molecular Formula C12H17ClO2
    Formula Weight 228.72
    MDL Number MFCD00048688
    Chemical Name or Material 2-Chloro-1,4-di-n-propoxybenzene

    Specifications

    Formula Weight 228.72
    CAS 916791-98-5
    Molecular Formula C12H17ClO2
    MDL Number MFCD00048688
    Quantity 10g
    Chemical Name or Material 2-Chloro-1,4-di-n-propoxybenzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit