Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-1,4-di-n-propoxybenzene, 97%

Click to view available options
Quantity:
10g
50g
250g
916791-98-5
C12H17ClO2
228.716
MFCD00048688
SWQFWWUXVCCQSJ-UHFFFAOYSA-N
2-chloro-1,4-di-n-propoxybenzene,chlorohydroquinone di-n-propyl ether,1-chloro-2,5-di-n-propoxybenzene
7020403
2-chloro-1,4-dipropoxybenzene
CCCOC1=CC(=C(C=C1)OCCC)Cl
916791-98-5
C12H17ClO2
228.716
MFCD00048688
SWQFWWUXVCCQSJ-UHFFFAOYSA-N
2-chloro-1,4-di-n-propoxybenzene,chlorohydroquinone di-n-propyl ether,1-chloro-2,5-di-n-propoxybenzene
7020403
2-chloro-1,4-dipropoxybenzene
CCCOC1=CC(=C(C=C1)OCCC)Cl
Quantity 50g
Formula Weight 228.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-1,4-di-n-propoxybenzene