Alfa Aesar™ 2-Chloro-1-(ethoxymethyl)imidazole, 97%

Manufacturer: Alfa Aesar™ H51870MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51870MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 850429-55-9
    Molecular Formula C6H9ClN2O
    Formula Weight 160.6
    MDL Number MFCD03788569
    Chemical Name or Material 2-Chloro-1-(ethoxymethyl)imidazole

    Specifications

    Formula Weight 160.6
    Molecular Formula C6H9ClN2O
    MDL Number MFCD03788569
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Quantity 250mg
    CAS 850429-55-9
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Chloro-1-(ethoxymethyl)imidazole
    Assay 97%

    Description

    Applications
    2-Chloro-1-(ethoxymethyl)imidazole is used as a pharmaceutical intermediate. It is also used in the synthesis of a new class of antifungal agents. Further, it is employed in Suzuki-Miyaura coupling reactions.

    Notes
    Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit