Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-1-fluoro-4-nitrobenzene, 98%

Click to view available options
Quantity:
25g
50g
500g
350-30-1
C6H3ClFNO2
175.543
MFCD00007206
DPHCXXYPSYMICK-UHFFFAOYSA-N
3-chloro-4-fluoronitrobenzene,benzene, 2-chloro-1-fluoro-4-nitro,3-chloro-4-fluoro-1-nitrobenzene,4-fluoro-3-chloronitrobenzene,1-chloro-2-fluoro-5-nitrobenzene,2-chloro-1-fluoro-4-nitro-benzene,3-chloro-4-fluoro-nitrobenzene,pubchem8539,acmc-209icj,dsstox_cid_29134
67688
2-chloro-1-fluoro-4-nitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])Cl)F
350-30-1
C6H3ClFNO2
175.543
MFCD00007206
DPHCXXYPSYMICK-UHFFFAOYSA-N
3-chloro-4-fluoronitrobenzene,benzene, 2-chloro-1-fluoro-4-nitro,3-chloro-4-fluoro-1-nitrobenzene,4-fluoro-3-chloronitrobenzene,1-chloro-2-fluoro-5-nitrobenzene,2-chloro-1-fluoro-4-nitro-benzene,3-chloro-4-fluoro-nitrobenzene,pubchem8539,acmc-209icj,dsstox_cid_29134
67688
2-chloro-1-fluoro-4-nitrobenzene
C1=CC(=C(C=C1[N+](=O)[O-])Cl)F
Density 1.61
Boiling Point 227°C to 232°C
Quantity 50g
Flash Point 112°C (233°F)
Melting Point 42°C to 45°C
Odor Characteristic
UN Number UN2811
Beilstein 1944997
Formula Weight 175.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-1-fluoro-4-nitrobenzene
Show More Show Less

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-499-3

RTECSNumber : CZ0720000

TSCA : No

Recommended Storage : Ambient temperatures